Assay Details
Target Gene Details
Entrez Gene ID: | 52882 |
Gene Name: | regulator of G-protein signalling 7 binding protein |
Gene Aliases: |
A930030I01Rik, AI842293, D13Bwg1146e, R7bp |
Location: |
Chr.13:104945904-105054930 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs7bp | NM_029879.2 | 6 | 2878 | NP_084155.2 |
AK028323.1 | 6 | 2255 | ||
AK036240.1 | 6 | 2786 | BAC29358.1 | |
AK043529.1 | 3 | 1682 |