Assay Details
Target Gene Details
Entrez Gene ID: | 11549 |
Gene Name: | adrenergic receptor, alpha 1a |
Gene Aliases: |
Adra1c |
Location: |
Chr.14:66635017-66771240 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adra1a | NM_001271759.1 | 4 | 3394 | NP_001258688.1 |
NM_001271760.1 | 4 | 3090 | NP_001258689.1 | |
NM_001271761.1 | NP_001258690.1 | |||
NM_013461.4 | 4 | 3553 | NP_038489.2 | |
AK033490.1 | 1 | 938 | BAC28316.1 | |
AK042759.1 | BAC31355.1 | |||
AK079597.1 | 4 | 3546 | BAC37694.1 | |
AK085653.1 | 4 | 3396 | BAC39495.1 | |
BC113139.1 | AAI13140.1 |
More Information
Set Membership: |
Intragenic Exonic |