Assay Details
Target Gene Details
Entrez Gene ID: | 328232 |
Gene Name: | glucose-fructose oxidoreductase domain containing 1 |
Gene Aliases: |
9630032O13, AI850995 |
Location: |
Chr.13:43195240-43304172 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfod1 | NM_001033399.4 | 2 | 4750 | NP_001028571.1 |
AK036079.1 | 1 | 2416 | ||
AK047231.1 | 1 | 450 | ||
AK050062.1 | 1 | 1535 | ||
AK147458.1 | 2 | 4751 | BAE27925.1 |
More Information
Set Membership: |
Intragenic Exonic |