Assay Details
Target Gene Details
Entrez Gene ID: | 27386 |
Gene Name: | neuronal PAS domain protein 3 |
Gene Aliases: |
4930423H22Rik, bHLHe12 |
Location: |
Chr.12:53248003-54072175 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npas3 | NM_013780.2 | 12 | 4396 | NP_038808.2 |
AK034372.1 | 1 | 512 | ||
AK162871.1 | 1 | 680 | BAE37094.1 |
More Information
Set Membership: |
Intragenic Exonic |