Assay Details
Target Gene Details
Entrez Gene ID: | 68680 |
Gene Name: | fat storage-inducing transmembrane protein 1 |
Gene Aliases: |
1110028A07Rik, AV010547, Cg10671, Fit1 |
Location: |
Chr.14:55575674-55576952 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fitm1 | NM_026808.1 | 1 | 128 | NP_081084.1 |
AB054000.1 | 1 | 128 | BAB47400.1 | |
AK003939.1 | BAB23085.1 | |||
BC037188.1 | 1 | 149 | AAH37188.1 | |
BC104408.1 | 1 | 128 | AAI04409.1 | |
BC104409.2 | 1 | 128 | AAI04410.1 |