Assay Details
Target Gene Details
Entrez Gene ID: | 66410 |
Gene Name: | mitochondrial transcription termination factor 3 |
Gene Aliases: |
2410017I18Rik, Mterfd1 |
Location: |
Chr.13:66905456-66933072 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mterf3 | NM_025547.3 | NP_079823.2 | ||
AK002299.1 | ||||
AK010525.1 | BAB27006.1 | |||
AK011934.1 | BAB27924.3 | |||
AK034645.1 | ||||
BC025173.1 | AAH25173.1 | |||
BC085282.1 | AAH85282.1 | |||
BY269653.1 |
Target Gene Details
Entrez Gene ID: | 19210 |
Gene Name: | phosphatidylserine synthase 1 |
Gene Aliases: |
AU044268, AW539008, PSS-1, mKIAA0024 |
Location: |
Chr.13:66932830-66998401 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptdss1 | NM_008959.3 | 1 | 150 | NP_032985.2 |
AK085857.1 | 1 | 150 | BAC39555.1 | |
AK172873.1 | 1 | 127 | BAD32151.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |