Assay Details
Target Gene Details
Entrez Gene ID: | 14538 |
Gene Name: | glucosaminyl (N-acetyl) transferase 2, I-branching enzyme |
Gene Aliases: |
5330430K10Rik, IGnT, IGnTA, IGnTB, IGnTC |
Location: |
Chr.13:40859768-40960892 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gcnt2 | NM_008105.3 | 3 | 3906 | NP_032131.2 |
NM_023887.4 | 3 | 3884 | NP_076376.3 | |
NM_133219.1 | 3 | 3546 | NP_573482.1 | |
AK077598.1 | 3 | 3521 | BAC36889.1 | |
AK080061.1 | 3 | 3473 | ||
AK146638.1 | 3 | 2648 | BAE27322.1 | |
AK146679.1 | 3 | 2648 | BAE27354.1 | |
AK156328.1 | 3 | 3420 | BAE33677.1 | |
AK169167.1 | 4 | 2694 | BAE40946.1 | |
AY435148.1 | 3 | 3297 | AAR95649.1 | |
AY435149.1 | 3 | 3297 | AAR95650.1 | |
AY435150.1 | 3 | 3297 | AAR95651.1 | |
BC054845.1 | 3 | 3528 | AAH54845.1 | |
BC094572.1 | 3 | 3884 | AAH94572.1 |
More Information
Set Membership: |
Intragenic Exonic |