Assay Details
Target Gene Details
Entrez Gene ID: | 15375 |
Gene Name: | forkhead box A1 |
Gene Aliases: |
Hnf-3a, Hnf3a, Tcf-3a, Tcf3a |
Location: |
Chr.12:57540628-57546911 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxa1 | NM_008259.3 | 1 | 389 | NP_032285.2 |
AB231452.1 | ||||
BC096524.1 | 1 | 389 | AAH96524.1 | |
U44752.1 | 1 | 230 | AAA86760.1 | |
X74936.1 | 1 | 196 | CAA52890.1 |
More Information
Set Membership: |
Intragenic Exonic |