Assay Details
Target Gene Details
Entrez Gene ID: | 105501 |
Gene Name: | abhydrolase domain containing 4 |
Gene Aliases: |
1110035H23Rik, AI429574, Abh4 |
Location: |
Chr.14:54254129-54269169 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abhd4 | NM_001205181.1 | 7 | 2424 | NP_001192110.1 |
NM_134076.2 | 7 | 2169 | NP_598837.2 | |
AK004112.1 | 1 | 1046 | ||
AK030250.1 | 7 | 2365 | BAC26863.1 | |
AK049366.1 | 7 | 2150 | BAC33711.1 | |
AK152595.1 | 7 | 2170 | BAE31344.1 | |
AK161853.1 | 7 | 2119 | BAE36606.1 | |
BC003982.1 | 5 | 1809 | AAH03982.1 | |
BC017532.1 | 7 | 2104 | AAH17532.1 |