Assay Details
Target Gene Details
Entrez Gene ID: | 14780 |
Gene Name: | glutathione peroxidase 5 |
Gene Aliases: |
AV379049, Arep |
Location: |
Chr.13:21286429-21292729 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpx5 | NM_010343.2 | 5 | 1440 | NP_034473.2 |
AK033841.1 | 2 | 1033 | BAC28492.1 | |
X53780.1 | 4 | 1276 | CAA37796.1 |
More Information
Set Membership: |
Intragenic Exonic |