Assay Details
Target Gene Details
Entrez Gene ID: | 14165 |
Gene Name: | fibroblast growth factor 10 |
Gene Aliases: |
AEY17, BB213776, Fgf-10, Gsfaey17 |
Location: |
Chr.13:118714699-118792573 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgf10 | NM_008002.4 | 3 | 3991 | NP_032028.1 |
AK141633.1 | 3 | 3313 | BAE24777.1 | |
BB487015.2 | 1 | 91 | ||
BC048229.1 | 3 | 3656 | AAH48229.1 |
More Information
Set Membership: |
Intragenic Exonic |