Assay Details
Target Gene Details
Entrez Gene ID: | 17977 |
Gene Name: | nuclear receptor coactivator 1 |
Gene Aliases: |
KAT13A, SRC-1, SRC1, bHLHe74 |
Location: |
Chr.12:4247362-4477210 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncoa1 | NM_010881.2 | 18 | 4374 | NP_035011.1 |
AK035922.1 | 3 | 698 | BAC29244.1 | |
BC068177.1 | 15 | 3682 | AAH68177.1 | |
BC080866.1 | 18 | 4137 | AAH80866.1 | |
U56920.1 | 17 | 3932 | AAB01228.1 | |
U64606.1 | 15 | 3677 | AAB06177.1 | |
U64828.1 | 18 | 4287 | AAB38841.1 |
More Information
Set Membership: |
Intragenic Exonic |