Assay Details
Target Gene Details
Entrez Gene ID: | 114712 |
Gene Name: | Fer3-like (Drosophila) |
Gene Aliases: |
Mnato3, N-twist, Nato, Nato3, Ntwist, bHLHa31, fer3 |
Location: |
Chr.12:33928425-33929310 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ferd3l | NM_033522.2 | 1 | 146 | NP_277057.1 |
AF517121.1 | 1 | 146 | AAN04085.1 | |
BC111576.1 | 1 | 138 | AAI11577.1 |
More Information
Set Membership: |
Intragenic Exonic |