Assay Details
Target Gene Details
Entrez Gene ID: | 105511 |
Gene Name: | family with sequence similarity 170, member B |
Gene Aliases: |
4922501K12Rik, AI449756 |
Location: |
Chr.14:32833962-32836789 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam170b | NM_001164485.1 | 2 | 898 | NP_001157957.1 |
AK076630.1 | 1 | 1674 | ||
BC117714.1 | 1 | 788 | ||
BC128503.1 | 1 | 788 | ||
BC128504.1 | 1 | 788 |