Assay Details
Target Gene Details
Entrez Gene ID: | 27277 |
Gene Name: | golgi autoantigen, golgin subfamily a, 5 |
Gene Aliases: |
Ret-II |
Location: |
Chr.12:102469134-102497911 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Golga5 | NM_001199004.1 | 13 | 2666 | NP_001185933.1 |
NM_013747.4 | 13 | 2508 | NP_038775.1 | |
AB016784.1 | 13 | 2667 | BAA33010.1 | |
AF026274.1 | 13 | 2498 | AAF21628.1 | |
AK138455.1 | BAE23668.1 | |||
AK152533.1 | 13 | 2516 | BAE31289.1 | |
AK153010.1 | 13 | 2516 | BAE31649.1 | |
AK168765.1 | 13 | 2512 | BAE40601.1 | |
BC016883.1 | 13 | 3236 | AAH16883.1 | |
BC086782.1 | 13 | 2508 | AAH86782.1 |