Assay Details
Target Gene Details
Entrez Gene ID: | 72308 |
Gene Name: | BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB (S. cerevisiae) |
Gene Aliases: |
2510002F24Rik, GTF3B, TAF3C, TAFIII90, TFIIIB90, mTFIIIB90 |
Location: |
Chr.12:112955631-113000707 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Brf1 | NM_028193.3 | 15 | 1955 | NP_082469.2 |
AK010890.1 | 2 | 204 | BAB27249.2 | |
BC037224.1 | 15 | 1738 | AAH37224.1 | |
BC058112.1 | 5 | 551 | AAH58112.1 |
More Information
Set Membership: |
Intragenic Exonic |