Assay Details
Target Gene Details
Entrez Gene ID: | 211922 |
Gene Name: | DENN/MADD domain containing 6A |
Gene Aliases: |
Fam116a |
Location: |
Chr.14:26573858-26634322 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dennd6a | NM_001134465.2 | 20 | 2219 | NP_001127937.1 |
NM_001285466.1 | 18 | 2065 | NP_001272395.1 | |
NM_001285467.1 | 20 | 2280 | NP_001272396.1 | |
AK033036.1 | 18 | 2067 | BAC28133.1 | |
AK050731.1 | 3 | 571 | ||
AK054197.1 | 20 | 2163 | BAC35690.1 | |
AK054295.1 | 20 | 2220 | BAC35721.1 | |
AK078910.1 | 4 | 718 | ||
AK080327.1 | 10 | 1160 | BAC37879.1 | |
AK131163.1 | 20 | 2191 | BAD21413.1 | |
AK132219.1 | 20 | 2216 | BAE21040.1 | |
AK135953.1 | 16 | 1855 | BAE22742.1 | |
AK163396.1 | 20 | 2279 | BAE37333.1 | |
BC031553.1 | 6 | 881 | AAH31553.1 |