Assay Details
Target Gene Details
Entrez Gene ID: | 66336 |
Gene Name: | centromere protein P |
Gene Aliases: |
1700022C02Rik, 4921518G09Rik |
Location: |
Chr.13:49464025-49652879 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cenpp | NM_025495.3 | 8 | 916 | NP_079771.1 |
AK006234.1 | 8 | 953 | BAB24472.1 | |
AK012863.1 | 8 | 920 | BAB28519.1 | |
BC147207.1 | 8 | 896 | ||
BC147208.1 | 8 | 896 |