Assay Details
Target Gene Details
Entrez Gene ID: | 238988 |
Gene Name: | ELKS/RAB6-interacting/CAST family member 2 |
Gene Aliases: |
6430531D06, CAST, CAST1/ERC2, D14Ertd171e, ELKS2alpha |
Location: |
Chr.14:27622410-28478537 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erc2 | NM_177814.4 | 16 | 5106 | NP_808482.2 |
AK051416.1 | 6 | 2598 | BAC34632.2 | |
AK122265.1 | 16 | 3896 | BAC65547.1 | |
AK147213.1 | 20 | 5259 | BAE27770.1 | |
BC056760.1 | 16 | 5106 | AAH56760.1 |
More Information
Set Membership: |
Intragenic Exonic |