Assay Details
Target Gene Details
Entrez Gene ID: | 218865 |
Gene Name: | choline dehydrogenase |
Gene Aliases: |
AU041472, D630034H06Rik |
Location: |
Chr.14:30009000-30040466 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chdh | NM_001136240.1 | 2 | 738 | NP_001129712.1 |
NM_172264.2 | 2 | 536 | NP_758468.2 | |
NM_175343.5 | 2 | 711 | NP_780552.1 | |
AK030900.1 | 2 | 712 | BAC27176.1 | |
AK052715.1 | 2 | 716 | ||
AK087570.1 | 2 | 646 | BAC39931.1 | |
AK164042.1 | 2 | 574 | BAE37601.1 | |
BC039186.1 | 2 | 536 | AAH39186.1 |
More Information
Set Membership: |
Intragenic Exonic |