Assay Details
Target Gene Details
Entrez Gene ID: | 20741 |
Gene Name: | spectrin beta, erythrocytic |
Gene Aliases: |
AI842465, D330027P03Rik, Gm1301, Spnb-1, Spnb1, ja, jaundiced, mKIAA4219 |
Location: |
Chr.12:76580488-76710547 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sptb | NM_013675.3 | 33 | 7096 | NP_038703.3 |
AK147299.1 | 33 | 7097 | BAE27831.1 | |
AK147684.1 | 33 | 6689 | BAE28073.1 | |
AK147704.1 | 33 | 6689 | BAE28085.1 | |
AK220551.1 | 14 | 2196 | BAD90537.1 | |
BC117711.2 | 16 | 2350 | ||
BC129806.1 | 32 | 6429 | ||
BC150783.1 | 32 | 6462 |