Assay Details
Target Gene Details
Entrez Gene ID: | 72400 |
Gene Name: | PIN2/TERF1 interacting, telomerase inhibitor 1 |
Gene Aliases: |
2210403I16Rik, 2610028A01Rik, 67-11-3, AU024023, LPTS, LPTS1 |
Location: |
Chr.14:63860301-63919862 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pinx1 | NM_028228.3 | 7 | 824 | NP_082504.1 |
AF321817.1 | 7 | 772 | AAL37221.1 | |
AF421879.1 | 7 | 707 | AAL32445.1 | |
AJ344106.1 | 6 | 596 | CAC51439.1 | |
AK011578.1 | 1 | 684 | ||
AK075840.1 | 5 | 594 | BAC35998.1 | |
BC115636.1 | 7 | 736 |
More Information
Set Membership: |
Intragenic Exonic |