Assay Details
Target Gene Details
Entrez Gene ID: | 12684 |
Gene Name: | cell death-inducing DNA fragmentation factor, alpha subunit-like effector B |
Gene Aliases: |
1110030C18Rik, AI790179, CIDE-B |
Location: |
Chr.14:55754052-55758424 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cideb | NM_009894.3 | 5 | 928 | NP_034024.2 |
AF041377.1 | 5 | 916 | AAC34986.1 | |
AK003968.1 | 5 | 963 | BAE43153.1 | |
BC012664.1 | 5 | 892 | AAH12664.1 | |
BC046340.1 | 5 | 922 | AAH46340.1 | |
BQ175754.1 | 1 | 259 |
Target Gene Details
Entrez Gene ID: | 67842 |
Gene Name: | NOP9 nucleolar protein |
Gene Aliases: |
2610027L16Rik |
Location: |
Chr.14:55745693-55755634 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nop9 | NM_026403.3 | 10 | 2494 | NP_080679.3 |
AK146687.1 | 10 | 2466 |
More Information
Set Membership: |
Intragenic Exonic |