Assay Details
Target Gene Details
Entrez Gene ID: | 13644 |
Gene Name: | embryonal Fyn-associated substrate |
Gene Aliases: |
- |
Location: |
Chr.14:54916543-54926788 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Efs | NM_010112.4 | 2 | 1536 | NP_034242.2 |
AK030321.1 | 2 | 663 | BAC26900.1 | |
AK163191.1 | 2 | 577 | BAE37228.1 | |
BC005438.1 | 2 | 605 | AAH05438.1 | |
BC031543.1 | 2 | 331 | AAH31543.1 | |
U28728.1 | 2 | 560 | AAC52340.1 | |
U57686.1 | 2 | 260 | AAB02246.1 |