Assay Details
Target Gene Details
Entrez Gene ID: | 108912 |
Gene Name: | cell division cycle associated 2 |
Gene Aliases: |
2610311M19Rik, AI586158 |
Location: |
Chr.14:67673180-67715841 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdca2 | NM_001110162.1 | 15 | 2446 | NP_001103632.1 |
NM_175384.4 | 15 | 2149 | NP_780593.3 | |
AK031313.1 | 11 | 1591 | BAC27343.1 | |
AK213197.1 | ||||
BC116298.1 | 11 | 1591 | ||
BC116299.1 | 11 | 1591 |