Assay Details
Target Gene Details
Entrez Gene ID: | 109700 |
Gene Name: | integrin alpha 1 |
Gene Aliases: |
CD49A, E130012M19Rik, Vla1 |
Location: |
Chr.13:114958079-115101964 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga1 | NM_001033228.3 | NP_001028400.2 | ||
AK029105.1 | ||||
AK035870.1 | BAE20498.1 | |||
AK043769.1 | BAE20643.1 | |||
AK137169.1 | ||||
AK141204.1 | BAE24585.1 | |||
AK165357.1 | ||||
AK169964.1 | ||||
BC014765.1 |
Target Gene Details
Entrez Gene ID: | 105083 |
Gene Name: | pelota homolog (Drosophila) |
Gene Aliases: |
AA409897, AI852218 |
Location: |
Chr.13:115088355-115090158 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pelo | NM_134058.3 | 2 | 1267 | NP_598819.2 |
AI854576.1 | 1 | 322 | ||
AK146382.1 | 2 | 1297 | BAE27128.1 | |
AK168228.1 | 2 | 1297 | BAE40181.1 | |
AK170635.1 | 2 | 1267 | BAE41926.1 | |
BB767039.1 | 1 | 242 | ||
BC050209.1 | 2 | 1180 | AAH50209.1 | |
BC057160.1 | 2 | 1068 | AAH57160.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |