Assay Details
Target Gene Details
Entrez Gene ID: | 22598 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 18 |
Gene Aliases: |
B0AT3, D630001K16Rik, Xt2, Xtrp2 |
Location: |
Chr.13:73661750-73678023 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc6a18 | NM_001040692.3 | 12 | 1840 | NP_001035782.1 |
NM_001136087.2 | 12 | 1726 | NP_001129559.1 | |
NM_001168643.1 | 11 | 1647 | NP_001162114.1 | |
NM_001168644.1 | 11 | 1677 | NP_001162115.1 | |
NM_001168645.1 | 11 | 1563 | NP_001162116.1 | |
NM_001168646.1 | 12 | 1810 | NP_001162117.1 | |
AF075262.1 | 12 | 1817 | AAC27757.1 | |
AF075263.1 | 12 | 1787 | AAC27758.1 | |
AF075264.1 | 12 | 1703 | AAC27759.1 | |
AF075265.1 | 11 | 1654 | AAC27760.1 | |
AF075266.1 | 11 | 1540 | AAC27761.1 | |
AF075267.1 | 12 | 1756 | AAC27762.1 | |
AK085258.1 | 11 | 1644 | ||
AK085473.1 | 12 | 1725 | ||
AK128907.1 | 12 | 1723 | ||
BC010748.1 | 12 | 1702 | AAH10748.1 |
More Information
Set Membership: |
Intragenic Exonic |