Assay Details
Target Gene Details
Entrez Gene ID: | 12161 |
Gene Name: | bone morphogenetic protein 6 |
Gene Aliases: |
D13Wsu115e, Vgr1 |
Location: |
Chr.13:38345716-38499728 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bmp6 | NM_007556.2 | 7 | 1737 | NP_031582.1 |
AK135770.1 | 7 | 1370 | BAE22651.1 | |
BC138593.1 | 7 | 1657 | ||
BC138595.1 | 7 | 1657 | ||
J04566.1 | 7 | 1319 | AAA40548.1 | |
X80992.1 | 7 | 1736 | CAA56917.1 |
More Information
Set Membership: |
Intragenic Exonic |