Assay Details
Target Gene Details
Entrez Gene ID: | 69480 |
Gene Name: | tetratricopeptide repeat domain 9 |
Gene Aliases: |
1700029M07Rik, AI429215, AI853559 |
Location: |
Chr.12:81629771-81664941 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc9 | NM_001033149.2 | 1 | 214 | NP_001028321.1 |
AK133456.1 | 1 | 215 | BAE21667.1 | |
AK161499.1 | BAE36425.1 | |||
BC028891.1 | 1 | 298 | AAH28891.1 | |
BC139242.1 | 1 | 212 | ||
BC139244.1 | 1 | 212 |
More Information
Set Membership: |
Intragenic Exonic |