Assay Details
Target Gene Details
Entrez Gene ID: | 76260 |
Gene Name: | tetratricopeptide repeat domain 8 |
Gene Aliases: |
0610012F22Rik, AV001447, BBS8 |
Location: |
Chr.12:98920574-98983250 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ttc8 | NM_029553.3 | 14 | 1747 | NP_083829.1 |
NM_198311.1 | 15 | 1739 | NP_938053.1 | |
AK002597.1 | 14 | 1733 | BAB22218.1 | |
AK032748.1 | 14 | 1709 | ||
AK081697.1 | 14 | 1714 | BAC38298.1 | |
BC017523.1 | 15 | 1739 | AAH17523.1 |