Assay Details
Target Gene Details
Entrez Gene ID: | 217946 |
Gene Name: | cell division cycle associated 7 like |
Gene Aliases: |
BC006933, JPO2 |
Location: |
Chr.12:117843475-117878712 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdca7l | NM_146040.1 | 10 | 2312 | NP_666152.1 |
AK149673.1 | 10 | 2290 | BAE29017.1 | |
BC006933.1 | 10 | 2312 | AAH06933.1 |
Target Gene Details
Entrez Gene ID: | 13411 |
Gene Name: | dynein, axonemal, heavy chain 11 |
Gene Aliases: |
Dnahc11, b2b1203Clo, b2b1279Clo, b2b1289Clo, b2b1727Clo, b2b598Clo, iv, lrd |
Location: |
Chr.12:117877982-118199043 on Build GRCm38 |
Assay Gene Location: | Within Exon 82 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dnah11 | NM_010060.3 | 82 | 13647 | NP_034190.3 |
AF183144.1 | 82 | 13647 | AAF07922.1 |
More Information
Set Membership: |
Intragenic Exonic |