Assay Details
Target Gene Details
Entrez Gene ID: | 14063 |
Gene Name: | coagulation factor II (thrombin) receptor-like 1 |
Gene Aliases: |
Gpcr11, PAR-2, Par2 |
Location: |
Chr.13:95511718-95525240 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
F2rl1 | NM_007974.4 | 2 | 1070 | NP_032000.3 |
AK144986.1 | 2 | 1054 | BAE26169.1 | |
AK158462.1 | 2 | 1058 | BAE34521.1 | |
AK160916.1 | 2 | 1058 | BAE36090.1 | |
AX785009.1 | 2 | 1029 | CAE11955.1 | |
BC025432.1 | 2 | 1055 | AAH25432.1 | |
Z48043.1 | 2 | 1029 | CAA88097.1 |
More Information
Set Membership: |
Intragenic Exonic |