Assay Details
Target Gene Details
Entrez Gene ID: | 13449 |
Gene Name: | docking protein 2 |
Gene Aliases: |
DokR, Frip |
Location: |
Chr.14:70764528-70778494 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dok2 | NM_010071.2 | 5 | 1537 | NP_034201.1 |
AF030627.1 | 5 | 1537 | AAC31315.1 | |
AF035117.1 | 5 | 1533 | AAC13266.1 | |
AF059583.1 | 5 | 1523 | AAC78606.1 | |
AK155579.1 | 5 | 1534 | BAE33334.1 | |
AK158246.1 | 5 | 1501 | BAE34423.1 | |
AK159466.1 | 5 | 1578 | BAE35107.1 | |
AK159472.1 | 5 | 1615 | BAE35113.1 | |
BC004590.1 | 5 | 1529 | AAH04590.1 |