Assay Details
Target Gene Details
Entrez Gene ID: | 68750 |
Gene Name: | ras responsive element binding protein 1 |
Gene Aliases: |
1110037N09Rik, AA414966, B930013M22Rik |
Location: |
Chr.13:37778382-37952005 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rreb1 | NM_026830.2 | 1 | 148 | NP_081106.1 |
AK154980.1 | 1 | 148 | BAE32969.1 | |
BC080680.1 | AAH80680.1 | |||
BY205187.1 | 1 | 151 |