Assay Details
Target Gene Details
Entrez Gene ID: | 20716 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 3N |
Gene Aliases: |
Spi2-2, Spi2.2, Spi2/eb.4 |
Location: |
Chr.12:104406675-104414334 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpina3n | NM_009252.2 | 3 | 837 | NP_033278.2 |
BC013651.1 | 3 | 816 | AAH13651.1 | |
M64086.1 | 3 | 792 | AAA40130.1 |
More Information
Set Membership: |
Intragenic Exonic |