Assay Details
Target Gene Details
Entrez Gene ID: | 20714 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 3K |
Gene Aliases: |
1300001I07Rik, D12Rp54, MMCM2, MMSpi2, RP54, Spi-2, Spi2, contrapsin |
Location: |
Chr.12:104338486-104345739 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpina3k | NM_011458.2 | 2 | 172 | NP_035588.2 |
AK004842.1 | 2 | 170 | ||
AK149436.1 | 2 | 173 | BAE28874.1 | |
BC011217.1 | 2 | 149 | AAH11217.1 | |
BC016407.1 | 2 | 160 | AAH16407.1 | |
BC019802.1 | 2 | 149 | AAH19802.1 | |
X55147.1 | 2 | 155 | CAA38948.1 | |
X56786.1 | 2 | 161 | CAA40106.1 |
More Information
Set Membership: |
Intragenic Exonic |