Assay Details
Target Gene Details
Entrez Gene ID: | 80898 |
Gene Name: | endoplasmic reticulum aminopeptidase 1 |
Gene Aliases: |
Arts1, ERAAP, PILSA, PILSAP |
Location: |
Chr.13:74639624-74691931 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erap1 | NM_030711.4 | 19 | 3151 | NP_109636.1 |
AK030329.1 | 19 | 3152 | BAC26904.1 | |
AK040350.1 | 16 | 2369 | BAC30569.1 |
More Information
Set Membership: |
Intragenic Exonic |