Assay Details
Target Gene Details
Entrez Gene ID: | 13803 |
Gene Name: | ectodermal-neural cortex 1 |
Gene Aliases: |
Nrpb, PIG10 |
Location: |
Chr.13:97241105-97253040 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Enc1 | NM_007930.4 | 2 | 729 | NP_031956.3 |
AK008709.1 | 2 | 545 | ||
AK044906.1 | 2 | 729 | BAC32136.1 | |
AK049187.1 | 2 | 724 | BAC33597.1 | |
AK132173.1 | 2 | 729 | BAE21011.1 | |
BC049186.1 | 2 | 360 | AAH49186.1 | |
BC058098.1 | 2 | 664 | AAH58098.1 | |
U65079.1 | 2 | 581 | AAB64206.1 |