Assay Details
Target Gene Details
Entrez Gene ID: | 20787 |
Gene Name: | sterol regulatory element binding transcription factor 1 |
Gene Aliases: |
ADD-1, ADD1, D630008H06, SREBP-1, SREBP-1a, SREBP-1c, SREBP1, SREBP1c, bHLHd1 |
Location: |
Chr.11:60199089-60222686 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Srebf1 | NM_011480.3 | 10 | 1997 | NP_035610.1 |
AF374266.1 | 9 | 1761 | AAK54762.1 | |
AK052628.1 | 10 | 1998 | BAC35068.1 | |
AK150052.1 | 10 | 1986 | BAE29268.1 | |
AK154424.1 | 10 | 1965 | BAE32576.1 | |
AK158269.1 | 3 | 546 | BAE34440.1 | |
AK169607.1 | 10 | 1998 | BAE41256.1 | |
BC006051.1 | 7 | 1133 | AAH06051.1 | |
BC056922.1 | 10 | 1979 | AAH56922.1 |