Assay Details
Target Gene Details
Entrez Gene ID: | 19364 |
Gene Name: | RAD51 homolog D |
Gene Aliases: |
R51H3, Rad51l3, TRAD |
Location: |
Chr.11:82871960-82890751 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rad51d | NM_001277938.1 | 10 | 3085 | NP_001264867.1 |
NM_001277939.1 | NP_001264868.1 | |||
NM_001277941.1 | 9 | 2965 | NP_001264870.1 | |
NM_001277942.1 | 8 | 2938 | NP_001264871.1 | |
NM_011235.4 | 10 | 3100 | NP_035365.1 | |
NR_102717.1 | 9 | 3029 | ||
NR_102718.1 | 10 | 3123 | ||
NR_102719.1 | 9 | 2981 | ||
NR_102720.1 | 9 | 2966 | ||
AK036272.1 | 9 | 2939 | ||
EU627688.1 |
More Information
Set Membership: |
Intragenic Exonic |