Assay Details
Target Gene Details
Entrez Gene ID: | 20185 |
Gene Name: | nuclear receptor co-repressor 1 |
Gene Aliases: |
5730405M06Rik, A230020K14Rik, N-CoR, RIP13, Rxrip13, mKIAA1047 |
Location: |
Chr.11:62315643-62457332 on Build GRCm38 |
Assay Gene Location: | Within Exon 38 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncor1 | NM_001252313.1 | 36 | 5759 | NP_001239242.1 |
NM_011308.3 | 36 | 5558 | NP_035438.3 | |
AB093281.1 | 18 | 3499 | BAC41465.1 | |
AK136008.1 | 1 | 126 | BAE22771.1 | |
AK137632.1 | 10 | 1877 | BAE23439.1 | |
BC086657.1 | 36 | 5524 | AAH86657.1 | |
L78294.1 | 15 | 2566 | AAL40135.1 | |
U22016.1 | 2 | 313 | AAC52168.1 | |
U35312.1 | 35 | 5802 | AAB17125.1 |
More Information
Set Membership: |
Intragenic Exonic |