Assay Details
Target Gene Details
Entrez Gene ID: | 67181 |
Gene Name: | CTD nuclear envelope phosphatase 1 |
Gene Aliases: |
2610507E10Rik, Dullard |
Location: |
Chr.11:69981168-69990601 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctdnep1 | NM_026017.2 | 1 | 65 | NP_080293.1 |
AK012063.1 | 1 | 49 | ||
AK076150.1 | 1 | 78 | ||
AK164602.1 | BAE37845.1 | |||
BC018265.1 | 1 | 65 | AAH18265.1 |
Target Gene Details
Entrez Gene ID: | 54351 |
Gene Name: | elongator acetyltransferase complex subunit 5 |
Gene Aliases: |
Derp6, Rai12 |
Location: |
Chr.11:69968224-69981294 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Elp5 | NM_018740.2 | NP_061210.2 | ||
AK152582.1 | BAE31332.1 | |||
AK217337.1 |