Assay Details
Target Gene Details
Entrez Gene ID: | 58172 |
Gene Name: | SERTA domain containing 2 |
Gene Aliases: |
AB041541, AU021878, Kiaa0127, MNCb-1504, SEI-2, Sei2, TRIP-Br2, mKIAA0127 |
Location: |
Chr.11:20542938-20653023 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sertad2 | NM_001038625.1 | 2 | 2903 | NP_001033714.1 |
NM_021372.2 | 2 | 3114 | NP_067347.2 | |
AK129059.1 | 1 | 2063 | BAC97869.1 | |
AK147674.1 | 3 | 3290 | BAE28066.1 | |
AK150033.1 | 1 | 518 |
More Information
Set Membership: |
Intragenic Exonic |