Assay Details
Target Gene Details
Entrez Gene ID: | 214384 |
Gene Name: | myocardin |
Gene Aliases: |
BSAC2A, Srfcp |
Location: |
Chr.11:65173608-65269989 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myocd | NM_145136.4 | 10 | 1600 | NP_660118.3 |
NM_146386.3 | 10 | 1600 | NP_666498.2 | |
AF384055.2 | 10 | 1566 | AAK71683.2 | |
AF437877.1 | 6 | 925 | AAL30892.1 | |
AK142216.1 | 10 | 1603 | BAE24980.1 | |
AY303755.1 | 10 | 1591 | AAQ63841.1 | |
BC141281.1 | 11 | 1646 |
More Information
Set Membership: |
Intragenic Exonic |