Assay Details
Target Gene Details
Entrez Gene ID: | 66213 |
Gene Name: | mediator complex subunit 7 |
Gene Aliases: |
1110063B05Rik, Crsp33, Crsp9 |
Location: |
Chr.11:46436933-46442721 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Med7 | NM_001104530.1 | 3 | 950 | NP_001098000.1 |
NM_001104556.1 | 2 | 948 | NP_001098026.1 | |
NM_001104557.1 | 2 | 780 | NP_001098027.1 | |
NM_025426.3 | 3 | 1118 | NP_079702.3 | |
AI844245.1 | 1 | 263 | ||
AK004354.1 | 3 | 1083 | BAB23272.1 | |
AK010357.1 | 3 | 946 | BAB26877.1 | |
AK012799.1 | 3 | 1098 | BAB28478.1 | |
AK144918.1 | 2 | 906 | BAE26135.1 | |
AK147739.1 | 3 | 1108 | BAE28106.1 | |
AK167407.1 | 3 | 1078 | BAE39496.1 | |
AK210054.1 | 1 | 81 | ||
BC011333.1 | 3 | 1105 | AAH11333.1 | |
CF983034.1 | 1 | 112 | ||
DV649959.1 | 1 | 87 |
More Information
Set Membership: |
Intragenic Exonic |