Assay Details
Target Gene Details
Entrez Gene ID: | 83813 |
Gene Name: | tyrosine kinase, non-receptor, 1 |
Gene Aliases: |
Kos1, Tnk1a, Tnk1b |
Location: |
Chr.11:69850563-69858730 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tnk1 | NM_031880.3 | 7 | 810 | NP_114086.3 |
AF307746.1 | 7 | 810 | AAL09413.1 | |
AF332512.1 | 5 | 630 | AAK35164.1 | |
AK033440.1 | 2 | 181 | BAC28288.1 | |
AK076220.1 | 7 | 817 | ||
AK167292.1 | 7 | 811 | BAE39397.1 | |
BC029623.1 | 6 | 797 | AAH29623.1 | |
BC055303.1 | 6 | 714 | AAH55303.1 |