Assay Details
Target Gene Details
Entrez Gene ID: | 66753 |
Gene Name: | endoplasmic reticulum lectin 1 |
Gene Aliases: |
4933407N01Rik |
Location: |
Chr.11:30865632-30954173 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erlec1 |
Target Gene Details
Entrez Gene ID: | 103554 |
Gene Name: | proteasome (prosome, macropain) activator subunit 4 |
Gene Aliases: |
AA409398, AU041366, TEMO, mKIAA0077 |
Location: |
Chr.11:30771328-30881150 on Build GRCm38 |
Assay Gene Location: | Within Exon 47 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psme4 | NM_134013.3 | 47 | 6201 | NP_598774.2 |
AK129049.1 | 6 | 4180 | BAC97859.1 | |
AK169447.1 | 6 | 1098 | BAE41181.1 | |
BC004575.1 | 5 | 1030 | AAH04575.1 | |
BC031174.1 | 19 | 2645 | AAH31174.1 | |
BC054364.1 | 5 | 947 | AAH54364.1 | |
BE944080.1 | 1 | 254 |