Assay Details
Target Gene Details
Entrez Gene ID: | 217303 |
Gene Name: | CD300A antigen |
Gene Aliases: |
B230315M08Rik, Clm8, LMIR1, MAIR-I, MAIR-Ia, MMAC8, Pigr4, mcpir1 |
Location: |
Chr.11:114889149-114904651 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd300a | NM_170758.3 | 1 | 137 | NP_739564.1 |
AK045869.1 | 1 | 137 | BAC32515.1 | |
BC139296.1 | 1 | 73 | ||
BC139297.1 | 1 | 73 |
More Information
Set Membership: |
Intragenic Exonic |