Assay Details
Target Gene Details
Entrez Gene ID: | 74136 |
Gene Name: | SEC14-like 1 (S. cerevisiae) |
Gene Aliases: |
1200017E04Rik, 2810012L19Rik, Naa-35 |
Location: |
Chr.11:117115172-117159268 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sec14l1 | NM_001166506.1 | 16 | 4363 | NP_001159978.1 |
NM_001166507.1 | 17 | 4007 | NP_001159979.1 | |
NM_028777.3 | 17 | 2512 | NP_083053.2 | |
AB185284.1 | 1 | 66 | ||
AK004818.1 | 17 | 2437 | BAB23589.1 | |
AK012734.1 | 1 | 523 | ||
AK172947.1 | 13 | 3898 | BAD32225.1 |