Assay Details
Target Gene Details
Entrez Gene ID: | 23939 |
Gene Name: | mitogen-activated protein kinase 7 |
Gene Aliases: |
BMK-1, BMK1, ERK-5, ERK5, Erk5-T, PRKM7 |
Location: |
Chr.11:61488812-61494266 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mapk7 | NM_001291033.1 | 4 | 1441 | NP_001277962.1 |
NM_001291034.1 | 3 | 1516 | NP_001277963.1 | |
NM_001291035.1 | 3 | 1657 | NP_001277964.1 | |
NM_001291036.1 | 4 | 1506 | NP_001277965.1 | |
NM_001291037.1 | 3 | 1755 | NP_001277966.1 | |
NM_011841.2 | 4 | 1391 | NP_035971.1 | |
AB019373.1 | 4 | 1341 | BAA82039.1 | |
AF126159.1 | 3 | 1154 | AAD39394.1 | |
AF126160.1 | 3 | 1204 | AAD39395.1 | |
AF126161.1 | 3 | 1574 | AAD39396.1 | |
AK148119.1 | 4 | 1373 | BAE28357.1 | |
AK155187.1 | 4 | 1309 | BAE33103.1 | |
AY534740.1 | 4 | 1325 | AAS38576.1 | |
BC100398.1 | 3 | 1458 | AAI00399.1 |
More Information
Set Membership: |
Intragenic Exonic |